missing translation for 'onlineSavingsMsg'
Learn More

anti-EPAS1, SMAD3 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0419.1Set

Product Code. 16191344

  • 7970.00 NOK / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: EPAS1 rabbit purified polyclonal antibody (20μg); SMAD3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This item is not returnable. View return policy

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the EPAS1 protein, and the other against the SMAD3 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

EPAS1
Monoclonal/Polyclonal
EPAS1
EPAS1
1 Set
Primary
Antibody pair set content: 1. EPAS1 rabbit purified polyclonal antibody (20 ug) 2. SMAD3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
ECYT4/HIF2A/HLF/MOP2/PASD2/bHLHe73
Mouse, Rabbit
RUO
2034/4088
Human
EPAS1 & SMAD3 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers