missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20303-1-AP-150UL
This item is not returnable.
View return policy
Description
ANKH is a multipass transmembrane protein that is expressed in joints and other tissues and controls pyrophosphate levels in cultured cells. Progressive ankylosis-mediated control of pyrophosphate levels has been suggested as a possible mechanism regulating tissue calcification and susceptibility to arthritis in higher animals.Specifications
ANKH | |
Polyclonal | |
Unconjugated | |
ANKH | |
ANK, ANKH, CCAL2, CMDJ, CPPDD, HANK, KIAA1581, MANK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56172 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HCJ1 | |
ANKH | |
ANKH Fusion Protein Ag14125 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction