Learn More
Hettich™ Angle Rotor ROTOFIX 32 A and Universal 320/320 R Centrifuge, 8-place
Ideal for routine laboratory tasks in doctors offices and small hospitals
Brand: Hettich™ 1418
This item is not returnable.
View return policy
Description
- Angle Rotor without adapters
- 8-place, 45°
- For use with Hettich Centrifuges
Specifications
Angle Rotor | |
4,500 rpm | |
122 mm | |
29 mm | |
45° |
3,305 x g | |
146 mm | |
107 mm | |
ROTOFIX 32 A and Universal 320/320 R Centrifuges | |
8 |