missing translation for 'onlineSavingsMsg'
Learn More

Angiotensin II, MP Biomedicals™

Angiotensin II Human From Synthetic, Powder

842.70 NOK - 2070.00 NOK

Specifications

CAS 68521-88-0
Color Off-white
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410912
View Documents
MP Biomedicals
0219505105
5 mg
842.70 NOK
5mg
Please to purchase this item. Need a web account? Register with us today!
11420912
View Documents
MP Biomedicals
0219505110
10 mg
2070.00 NOK
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Angiotensin II plays an important role in the regulation of blood pressure.
  • It stimulates the release of aldosterone from the adrenal gland.
  • Angiotensin II is a peptide hormone which plays an important role in the regulation of blood pressure.
  • It stimulates the release of aldosterone from the adrenal gland.It has strong vasoconstrictive effects.
Specifications

Specifications

68521-88-0
Off-white
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Angiotensin II, MP Biomedicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.