Learn More
Labconco™ Ampules
Labconco Ampules are fabricated of highest quality borosilicate glass for strength and durability.
4135.00 NOK - 8955.00 NOK
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | |||||
10024084
|
Labconco™
7571010 |
5 mL | Pre-scored, flat-bottom |
6440.00 NOK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
|||||
10084314
|
Labconco™
7570610 |
1 mL | Pre-scored, flat-bottom |
4135.00 NOK
Pack of 100 |
Estimated Shipment: 28-10-2025
Log in to see stock. |
|||||
10359744
|
Labconco™
7571210 |
10 mL | Pre-scored, flat-bottom |
7455.00 NOK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
|||||
15103065
|
Labconco™
7572710 |
1 mL | Straight-sided |
8955.00 NOK
100 ampoules |
Estimated Shipment: 24-11-2025
Log in to see stock. |
|||||
Description
- Constructed of high quality borosilicate glass for strength and durability
- Supplied in packages of 100
Specifications
Borosilicate Glass | |
Ampule |
Product Title
Labconco™ Ampules