missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AMPK alpha-1/beta-2/gamma-2, GST Tag, His Tag Recombinant Protein
Human AMPK alpha-1/beta-2/gamma-2, GST Tag, His Tag Recombinant Protein
Brand: Invitrogen™ A33720
This item is not returnable.
View return policy
Description
AMPK (A1/B2/G2) serine/threonine kinase or AMP-activated protein kinase (AMPK) exhibits a key role as a master regulator of cellular energy homeostasis. AMPK exists as a heterotrimeric complex composed of a catalytic alpha subunit and regulatory beta and gamma subunits. Binding of AMP to the gamma subunit allosterically activates the complex. AMPK is activated in response to stresses that deplete cellular ATP (low glucose, hypoxia and ischemia) and via signaling pathways in response to adiponectin, leptin and CAMKK beta.- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Specifications
PRKAA1: NP_006242.4, PRKAB2: NP_005390.1, PRKAG2: NP_001035723.1 | |
90.3/56.6/85.6 kDa | |
CAMKL Kinase Family | |
Human | |
GST Tag, His Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
1 mg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction