missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B, MP Biomedicals

1205.00 NOK - 24095.00 NOK

Chemical Identifiers

CAS 1397-89-3
Molecular Formula C47H73NO17
Molecular Weight (g/mol) 924.09
MDL Number MFCD00877763
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
Synonym Amphomoronal, Fungizone, Amphotericine B
PubChem CID 134129663
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17660917
View Documents
MP Biomedicals
0219504380
100 mg
1205.00 NOK
100mg
Please to purchase this item. Need a web account? Register with us today!
11420892
View Documents
MP Biomedicals
0219504383
250 mg
2095.00 NOK
250mg
Please to purchase this item. Need a web account? Register with us today!
11430892
View Documents
MP Biomedicals
0219504390
500 mg
3615.00 NOK
500mg
Please to purchase this item. Need a web account? Register with us today!
11440892
View Documents
MP Biomedicals
0219504391
1 g
6300.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
11450892
View Documents
MP Biomedicals
0219504394
5 g
24095.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amphotericin B, MP Biomedicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.