missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
CAS | 7783-20-2 |
Quantity | 500 g |
Molecular Formula | H8N2O4S |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
SMILES | N.N.OS(O)(=O)=O |
IUPAC Name | sulfuric acid diamine |
Molecular Weight (g/mol) | 132.13 |
Grade | Reagent |
Safety and Handling

Product Identifier
- ammonium sulphate, reagent grade, ACS, ISO, rea. Ph eur
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ammonium sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium sulfate, ExpertQ, Scharlab >
Spot an opportunity for improvement?Share a Content Correction