missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Dibasic, Reagent Grade, ≥98%, Honeywell™
Reagent Grade,≥98.0%
309.35 NOK - 2719.83 NOK
Chemical Identifiers
CAS | 7783-28-0 |
---|---|
Molecular Formula | H9N2O4P |
Molecular Weight (g/mol) | 132.06 |
MDL Number | MFCD00010891 |
InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
PubChem CID | 24540 |
ChEBI | CHEBI:63051 |
SMILES | N.N.OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15677650
|
Honeywell Research Chemicals
A5764-100G |
100 g | Poly Bottle | N/A | ||||||
15607660
|
Honeywell Research Chemicals
A5764-500G |
500 g | Poly Bottle | N/A | ||||||
15687650
|
Honeywell Research Chemicals
A5764-1KG |
1 kg | Poly Bottle | N/A | ||||||
15697650
|
Honeywell Research Chemicals
A5764-2.5KG |
2.5 kg | Poly Bottle | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7783-28-0 | |
132.06 | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
24540 | |
N.N.OP(O)(O)=O |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
CHEBI:63051 |
Specifications
155°C | |
White | |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
N.N.OP(O)(O)=O | |
132.06 | |
CHEBI:63051 | |
≥98% | |
Crystalline | |
Ammonium phosphate dibasic |
7783-28-0 | |
7.5 to 9 (at 25°C) | |
(NH4)2HPO4 | |
NONH for all modes of transport | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
diazanium;hydrogen phosphate | |
24540 | |
132.06g/mol | |
Odorless | |
1.62g/cm3 at 20°C |
Safety and Handling
missing translation for 'rtecsNumber' : 231-987-8
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Phosphate Dibasic, Reagent Grade, ≥98%, Honeywell™