missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Molecular Biology Reagent, MP Biomedicals™

Product Code. 11406421
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ammonium Persulfate, Molecular Biology Reagent is a polymerization catalyst used with TEMED for polyacrylamide gel formation.

  • Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation.
  • APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry.
  • Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, and the decolorizing and deodorizing of oils.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (Decomposition)
Color White
Physical Form Crystalline Powder
Quantity 25 g
Solubility Information Soluble in water, 83.5 g/100 g water at 25°C
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Formula Weight 228.18
Percent Purity ≥98%
Grade Molecular Biology
Density 1.98 g/cm3 at 68°F; Bulk Density: 951 to 1050 kg/m3
Chemical Name or Material Ammonium persulfate
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'