missing translation for 'onlineSavingsMsg'
Learn More

Ammonium O,O'-diethyl dithiophosphate, typically 95%, Thermo Scientific Chemicals

739.00 NOK - 1460.00 NOK

Chemical Identifiers

CAS 1068-22-0
Molecular Formula C4H14NO2PS2
Molecular Weight (g/mol) 203.255
MDL Number MFCD00012635
InChI Key HFRHTRKMBOQLLL-UHFFFAOYSA-N
Synonym diethyl dithiophosphate, ammonium salt, ammonium ethyl phosphorodithioate, diethyl dithiophosphate ammonium salt, ammonium o,o-diethyldithiophosphate, ammonium diethyl dithiophosphate, c4h11o2ps2.nh3, wln: shps&o2&o2 &zh, ammonium di-o-ethyl dithiophosphate, ammonium,o-diethyl phosphorodithioate, ammonium o,o'-diethyl dithiophosphate
PubChem CID 12614240
IUPAC Name azane;diethoxy-sulfanyl-sulfanylidene-$l^{5}-phosphane
SMILES CCOP(=S)(OCC)S.N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357228
SDS View Documents
Thermo Scientific Alfa Aesar
030139.06
5 g
739.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
11367228
SDS View Documents
Thermo Scientific Alfa Aesar
030139.14
25 g
1460.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Diethyl dithiophosphate ammonium salt can be used in the extraction of arsenic. It is used as a source of the (C2H5O)2PS2- ligand in coordination chemistry and in analytical chemistry for determination of various ions. It can be obtained by the reaction of phosphorus pentasulfide with ethanol and ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 213-942-4

missing translation for 'rtecsNumber' : BP8145000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium O,O'-diethyl dithiophosphate, typically 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.