missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Hydroxide, (ACS Reagent Grade), MP Biomedicals Product Code.: 11496241

Ammonium Hydroxide, (ACS Reagent Grade), MP Biomedicals

Product Code. 11496241 Shop All MP Biomedicals Products
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 11496241

Brand: MP Biomedicals 0219385490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1336-21-6
Quantity 500 mL
Molecular Formula H5NO
MDL Number MFCD00066650
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.1
Grade ACS Reagent
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Hydroxide, (ACS Reagent Grade), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.