missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Chloride, ACS Reagent Grade, Scharlab Product Code.: 11379833
compliance-icons

Ammonium Chloride, ACS Reagent Grade, Scharlab

Product Code. 11379833
500 g, 500g
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 11379833

Brand: Scharlab AM02730500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in the manufacture of dyes, in explosive compositions, and in analytical chemistry.

  • Synonyms: Salt ammoniac
  • Molecular Formula: NH4Cl
  • Molar Mass: 53.49g/mol
  • CAS: 12125-02-9
  • EIN ECS No.: 235-186-4

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumkloridShow MoreShow Less
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Melting Point 335°C (decomposition)
pH 4.5–5.5 (5%, water, at 25°C)
Quantity 500 g
Formula Weight 53.49g/mol
Percent Purity 99.5% min (argentometric)
Grade ACS Reagent
Ignition Point >400°C
Loss on Drying 1% max (105°C)
Residue after Ignition 0.01% max (as SO4)
Appearance Passes test
Vapor Pressure 1.3hPa at 30°C
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium chloride, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Chloride, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.