missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Carbonate, Certified AR for Analysis, Powder, Fisher Chemical™ Product Code.: 10408210

Ammonium Carbonate, Certified AR for Analysis, Powder, Fisher Chemical™

Product Code. p-7072808 Shop All Fisher Chemical Products
500 g, 500g
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10408210

Brand: Fisher Chemical A/3680/53

Spotlight
Use promo code 24156 at checkout
View Available Promotions
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
MDL Number 10890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
IUPAC Name carbonic acid diamine
SMILES N.N.OC(O)=O

Specifications

Melting Point 58°C
Color White
pH 9.4
Physical Form Solid
Quantity 500 g
Formula Weight 96.09g/mol
Packaging Plastic powder jar
Vapor Pressure 69 hPa at 20°C
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Carbonate, Certified AR for Analysis, Powder, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.