Learn More
Ammonia Solution, Chem Lab™ Product Code.: 11997656
Ammonia Solution, Chem Lab™
Product Code. 11997656
5 L, 5 L
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. 11997656
Brand: Chem Lab CL00.0103.5000
Specifications
Color Colorless
Boiling Point 38°C
Melting Point -57°C
Composition 250 g NH3 / kg
Density 0.91g/mL
Solubility Information Water Soluble
Odor Odorless
pH >10
Quantity 5 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia 25 weight % solution p.
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonia
Product Content Correction
Product Title
Ammonia Solution, Chem Lab™ >