Learn More
Ammonia Solution, 0.1M, Fisher Chemical™ Product Code.: 12990904
Ammonia Solution, 0.1M, Fisher Chemical™
Product Code. 12990904 Shop All Fisher Chemical Products
1000 mL
Click to view available options
Quantity:
1000 mL
Product Code. 12990904
Brand: Fisher Chemical VL 202 1000
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Concentration 0.1mol/L
Quantity 1000 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1078508 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 202 1000
Lot Number1046205 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 202 1000
Lot Number1023658 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 202 1000
Lot Number1048344 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 202 1000
Product Content Correction
Product Title
Ammonia Solution, 0.1M, Fisher Chemical™ >