missing translation for 'onlineSavingsMsg'
Learn More
AMG-3969, MedChemExpress Product Code.: 16487867

AMG-3969, MedChemExpress

Product Code. p-200099184
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487867

Brand: Medchem Express HY12411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMG-3969 is a potent glucokinase-glucokinase regulatory protein interaction (GK-GKRP) disruptor with an IC50 of 4 nM.

AMG-3969 is a potent glucokinase-glucokinase regulatory protein interaction (GK-GKRP) disruptor with an IC50 of 4 nM.

Specifications

Chemical Name or Material AMG-3969
CAS 1361224-53-4
Color White
Purity Grade Notes Research
Molecular Formula C21H20F6N4O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (191.40 mM)
SMILES CC#C[C@@H]1N(C2=CC=C(C(C(F)(F)F)(O)C(F)(F)F)C=C2)CCN(S(=O)(C3=CN=C(N)C=C3)=O)C1
Molecular Weight (g/mol) 522.46
Formula Weight 522.46
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
AMG-3969, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.