Learn More
Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System
Shop All Cytiva Products
Click to view available options
Quantity:
For 2000 cm2 Membrane
For 4000 cm2 membrane
Labels 10 μg Nucleic Acid
Labels 5 μg Nucleic Acid
Product Type:
Detection Reagents
Hybridization Buffer
Labeling Module
Labeling and Detection System
Unit Size:
5µg
Each
Description
Includes
45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
- Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
- Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
- No need to strip blots before reprobing.
- For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
- Consistent results combining strong signals with very low backgrounds.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Colony or plaque screens, dot blots and PCR product analyses |
| Includes | ECL Detection Reagents 1 and 2, for 2000cm2 membrane |
| Product Type | Detection Reagents |
| Quantity | For 2000 cm2 Membrane |
| Test Time | 20 min. |