Learn More
VITLAB™ Amber Glass Reservoir Bottle, Square
Click to view available options
Capacity (Metric):
1000 mL
Description
- Square
- Amber
Specifications
Specifications
Capacity (Metric) | 1000 mL |
Coating | Ethylene Acrylate |
Color | Amber |
For Use With (Application) | For volume measurement |
Includes | Screw cap |
Material | Glass |
Max. Temperature (Metric) | 80°C |
No. per Pack | 1Each |
Operating Temperature | 80°C |
Quantity | 1Each |
Show More |
Product Content Correction
Product Title
VITLAB™ Amber Glass Reservoir Bottle, Square >