Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 2
Description
- Manufactured from Boro 3.3 glass
- Acc. to DIN EN ISO 1042
- Includes USP batch certificate
- Class A, DE-M marking
Specifications
Specifications
| Closure Material | PP (Polypropylene) |
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass (3.3) |
| Product Type | Volumetric flask |
| Class | A |
| Closure Type | Stopper |