Learn More
AM211, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.
Specifications
Specifications
Chemical Name or Material | AM211 |
CAS | 1175526-27-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C27H27F3N2O4 |
Quantity | 10 mg |
Synonym | AM211 free acid |
Solubility Information | DMSO : ≥ 125 mg/mL (249.75 mM) |
SMILES | O=C(O)CC1=CC(C2=CC=C(C(F)(F)F)C=C2CN(CC)C(NCC3=CC=CC=C3)=O)=C(OC)C=C1 |
Molecular Weight (g/mol) | 500.51 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AM211, MedChemExpress