Learn More
AM211, MedChemExpress Product Code.: 16436116
AM211, MedChemExpress
Product Code. 16436116
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16436116
Brand: Medchem Express HY13213
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.
AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.
Specifications
Chemical Name or Material AM211
CAS 1175526-27-8
Color White
Purity Grade Notes Research
Molecular Formula C27H27F3N2O4
Quantity 100 mg
Synonym AM211 free acid
Solubility Information DMSO : ≥ 125 mg/mL (249.75 mM)
SMILES O=C(O)CC1=CC(C2=CC=C(C(F)(F)F)C=C2CN(CC)C(NCC3=CC=CC=C3)=O)=C(OC)C=C1
Molecular Weight (g/mol) 500.51
Formula Weight 500.51
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AM211, MedChemExpress