Learn More
AM095 free acid, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 ÎĽM for recombinant human or mouse LPA1 respectively.
Specifications
Specifications
Chemical Name or Material | AM095 free acid |
CAS | 1228690-36-5 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C27H24N2O5 |
Quantity | 500 mg |
Solubility Information | DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming) |
SMILES | O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1 |
Molecular Weight (g/mol) | 456.49 |
Formula Weight | 456.49 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AM095 free acid, MedChemExpress