Learn More
Korff™ Aluminum Sheets
Brand:  Korff™ 60280
Product Code. 10158881
  • 1435.00 NOK / Pack of 1000
Only null left
Estimated Shipment: 18-08-2025
Specifications
Sheets
140 mm
Aluminum
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Korff™ Aluminum Sheets > Width: 140mm