Learn More
Cole-Parmer™ Aluminum Scoop
Brand:  Cole-Parmer™ 07207-06
 View more versions of this product
Product Code. 11775168
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Scoop
Ribbed
9 in.
Aluminum
1100 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Aluminum Scoop > Capacity: 1100mL