Learn More
Lenz Laborglasinstrumente™ Aluminum Rings with Hooks
Constructed from aluminum. Lenz Laborglasinstrumente™ Aluminum Rings with Hooks are perfect for attaching coil springs and securing connections of conical sections. The rings prevent the breakage and rejection of glass hooks.
Brand:  Lenz Laborglasinstrumente™ 01102514
Product Code. 11993351
  • 723.00 NOK / Pack of 10
Only null left
Estimated Shipment: 28-07-2025
Description
  • Made from Aluminum
  • Prevents breakage and rejection of glass hooks
Specifications
Specifications
Aluminum
10
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Aluminum Rings with Hooks > Material: Aluminum