man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Film and Foil Wrapping and Accessories
Film and Foil Wrapping
Korff™ Aluminum Refill Rolls
10250452
Print
Korff™ Aluminum Refill Rolls
Width: 300mm
Brand:
Korff™ 60170
View more versions of this product
Product Code.
10250452
600.00 NOK
/ 10m
Quantity
Estimated Shipment: 08-11-2024
Log in
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
This item is not returnable.
View return policy
Specifications
Product Type
Refill Rolls
Width (Metric)
300mm
Material
Aluminum