Learn More
Korff™ Aluminum Professional Rolls
Brand:  Korff™ 60110
 View more versions of this product
Product Code. 10730193
  • 1055.00 NOK / 150m
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Specifications
Professional Rolls
450mm
Aluminum
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Korff™ Aluminum Professional Rolls > Width: 450mm; Foil Gauge: 0.015mm