Learn More
Korff™ Aluminum Professional Rolls
Brand:  Korff™ 60150
 View more versions of this product
Product Code. 10322232
  • 2075.00 NOK / 100m
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Specifications
Professional Rolls
600mm
Aluminum
Documents
One moment while we fetch your results.
Product Title
Korff™ Aluminum Professional Rolls > Width: 600mm; Foil Gauge: 0.030mm