Learn More
Korff™ Aluminum Foil
Brand:  Korff™ 60019
 View more versions of this product
Product Code. 10515585
  • 1925.00 NOK / 600m
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Specifications
150 mm
Aluminum
20 μm
Product Title
Korff™ Aluminum Foil > Size: 0.02 x 150mm