Learn More
RETSCH Aluminum Cup
Brand:  RETSCH 22.458.0006
Product Code. 11924762
  • 6950.00 NOK / Pack of 1000
Only null left
Estimated Shipment: 01-09-2025
Specifications
Aluminum Cup
40 mm
Aluminum
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Aluminum Cup > Dia.: 40mm