Learn More
VACUUBRAND™ Aluminum Clamping Rings
A rapid connection that can be mounted without any tools for soft seals and indium seals.
Brand:  VACUUBRAND™ 20660000
237.36 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 10419122
  • 268.00 NOK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
Estimated Shipment: 16-06-2025
Description
  • Provided with wing nut
  • Available in two different sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/16 mm
Aluminum
DN 10/16 mm
For Small KF Flange
1 Each
Clamping Ring
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
    Days
    70
    Hours
    12
    Minutes
    53
    Seconds
    8
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    15
    Hours
    12
    Minutes
    53
    Seconds
    8
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
VACUUBRAND™ Aluminum Clamping Rings > Size: DN 10/16mm