Learn More
Bochem™ Aluminum Burette Clamp
Brand:  Bochem™ 5622
Product Code. 10618752
  • 224.00 NOK / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Specifications
Aluminum
Buret Clamp
4 to 20 mm
Product Title
Bochem™ Aluminum Burette Clamp > Dia.: 13.5 mm