Learn More
Fisherbrand™ Aluminum Alloy Rods
Approximately 13mm in diameter.
Brand:  Fisherbrand™ STH-710-030U
 View more versions of this product
Product Code. 10265394
  • 40.35 NOK / Each
Only null left
Estimated Shipment: 13-08-2025
Add to basket
Description
Description
  • Available in a range of lengths
  • Excellent for building scaffolding in the laboratory
Specifications
Specifications
Aluminum Alloy
100 mm
Rod
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Aluminum Alloy Rods > Length: 100mm