Learn More
Fisherbrand™ Aluminum Alloy Rods
Approximately 13mm in diameter.
Brand:  Fisherbrand™ STH-710-050X
 View more versions of this product
Product Code. 10133784
  • 50.75 NOK / Each
Only null left
Estimated Shipment: 16-07-2025
Add to basket
Description
Description
  • Available in a range of lengths
  • Excellent for building scaffolding in the laboratory
Specifications
Specifications
Aluminum Alloy
200 mm
Rod
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Aluminum Alloy Rods > Length: 200mm