Learn More
Ansell AlphaTec™ 58-535B Series Nitrile Immersion Gloves
Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
6 pairs
Description
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals
- Chemical resistant
- Material fully coated nitrile gloves
- Thickness: 11 mil
- Embossed grip surface for superior wet and oil grip
- Red with black coating
- Comfortable design reduces fatigue
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR 177-2600 |
| Applications | Industrial |
| Color | Green, Gray |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Chemical-resistant Gloves |
| Size | 8 |
| Glove Cuff Style | Gauntlet |
| Liner Material | Acrylic |
| Quantity | 6 Pairs |
| Show More |