Description
- Designed to protect, rear entry double zip system, attached socks with boot overflap
- Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Attached Ansell Barrier™ gloves, exhalation valves and clear face visor
Specifications
Specifications
Color | Green |
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
Material | Multi-Layer Non-Woven Barrier Laminate Fabric |
Size | 3X-Large |
Seam Type | Welded and Taped |
Style | G40T126B |
Weight (Metric) | 103 gsm |
Product Content Correction
Product Title
Ansell AlphaTec™ 4000 APOLLO >