Learn More
Ansell AlphaTec™ 38001PP Medium Duty Chemical-resistant Gloves with Gauntlet Cuff
Shop All Ansell Products
missing translation for 'orderingAttributeHoverText'
Size:
7
8
9
10
11
Description
- High flexibility and tactility performance vs. standard butyl
- Raised diamond pattern for enhanced grip
- High performance against highly aggressive chemicals including solvents, aldehydes, ketones, esters and concentrated mineral acids
Specifications
Specifications
Applications | Industrial |
Color | Black |
Latex Free | Yes |
Material | Butyl |
Size | 7 |
Thickness (English) | 14 mil |
Thickness (Metric) | 0.35 mm |
Glove Cuff Style | Gauntlet |
Length (English) | 14 in. |
Product Content Correction
Product Title
Ansell AlphaTec™ 38001PP Medium Duty Chemical-resistant Gloves with Gauntlet Cuff >