missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1500 PLUS Coverall with Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Proven protection, filters 99.9% of particles >3 microns
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Silicone free, critical in spray painting applications
- Anti-static
Specifications
Specifications
Size | Medium |
Material | Anti-static SMS |
Color | White, Blue, Navy, Orange |
Seam Type | Stitched |
Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2018, EN ISO 13688:2013 |
For Use With (Application) | Unpacking of Bulk Goods, Loading and Unloading of Proces Equipment, Body Surface Checking, Marking and Touching up, Assembly & Inspection of Components, Painting or Cleaning Tools or Robots, Production Line Support & Maintenance |
Weight (Metric) | 48 gsm |
Style | N15S111 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction