Learn More
Ansell AlphaTec™ 1500 PLUS Coverall with Hood
Product Code. 16834680
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Anti-static suit, used for the stripping, clear up or handling of asbestos, general maintenance, construction and contract cleaning.
  • Proven protection, filters 99.9% of particles >3 microns
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Silicone free, critical in spray painting applications
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Anti-static SMS
Color White, Blue, Navy, Orange
Seam Type Stitched
Certifications/Compliance (Europe) EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2018, EN ISO 13688:2013
For Use With (Application) Unpacking of Bulk Goods, Loading and Unloading of Proces Equipment, Body Surface Checking, Marking and Touching up, Assembly & Inspection of Components, Painting or Cleaning Tools or Robots, Production Line Support & Maintenance
Weight (Metric) 48 gsm
Style N15S111