missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1500 Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
B15S113
N15T138
O15S113
O15S138
R15S113
R15T138
W15T138
Description
- 3 piece hood
- Elasticated hood, wrists, waist, and ankles
- 2-way front zipper with resealable storm flap
- Finger loops
Specifications
Specifications
Size | Large |
Material | SMS Fabric |
Color | Red, Orange |
Seam Type | Stitched |
Certifications/Compliance (Europe) | EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN ISO 13688:2013 |
For Use With (Application) | Inspection (Picking, Checking and Marking), Asbestos Related Work, Handling of Powders |
Weight (Metric) | Red 50 gsm, Orange 50 gsm |
Style | B15S113 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction