Learn More
Abnova™ ALKBH5 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Brand: Abnova™ H00054890-T02.100uL
This item is not returnable.
View return policy
Specifications
293T | |
ALKBH5 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 51.4kDa | |
Transient overexpression cell lysate was tested with anti-ALKBH5 antibody (Mfr. No. H00054890-B01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
NM_017758 | |
pCMV-ALKBH5 full-length | |
100 μL | |
Human |