Learn More
3M™ Air Supply Tubes
Click to view available options
:
Length: 10m; For use with S-200 Supplied Air Respiratory System
Specifications
Specifications
Color | Blue |
Product Type | Air Supply Tube |
For Use With (Equipment) | To fit S-200 Supplied Air Respiratory System |
Length (Metric) | 10 m |
Product Content Correction
Product Title
3M™ Air Supply Tubes >