missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHCYL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | AHCYL1 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897173
|
Proteintech
10658-3-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887173
|
Proteintech
10658-3-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AHCYL1 belongs to the adenosylhomocysteinase family. The protein, an inositol 1, 4, 5-trisphosphate receptor-binding protein, specifically binds to and activates pancreas-type Na+/HCO3- cotransporter 1 (pNBC1). The regulation through AHCYL1 enables NBC1 variants to have different physiological roles.Specifications
AHCYL1 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
B5DFN2, O43865, Q80SW1 | |
10768, 229709, 362013 | |
AHCYL1 Fusion Protein Ag1017 | |
Primary | |
-20°C | |
AHCYL1 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AdoHcyase 2, AHCYL1, DCAL, IRBIT, PRO0233, XPVKONA | |
Ahcyl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title