missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPAT6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16762-1-AP-150UL
This item is not returnable.
View return policy
Description
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.Specifications
AGPAT6 | |
Polyclonal | |
Unconjugated | |
GPAT4 | |
1 AGP acyltransferase 6, 1 AGPAT 6, AGPAT6, GPAT4, LPAAT zeta, LPAATZ, TSARG7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102247, 137964, 290843 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86UL3, Q8K2C8 | |
Gpat4 | |
AGPAT6 Fusion Protein Ag10294 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction