Learn More
BTX™ AgilePulse™ MAX Chamber Holder for Large Volume 5 mL Chambers
AgilePulse™ MAX Chamber Holder for Large Volume 5 mL Chambers for use with AgilePulse™ MAX.
Brand: BTX™ 470202N
This item is not returnable.
View return policy
Description
- cuvette module plugs directly into generator
- Holds one cuvette
- Safety swing shield secures cuvette in holder
Specifications
Large Volume Electroporation | |
Chamber Holder | |
AgilePulse MAX Chamber Holder for Large Volume 5 ml Chambers | |
Chamber Holder Only | |
BNC Cable | |
Square Wave |
AgilePulse™ MAX, ECM™ 2001+ | |
50 to 1000 V | |
Touchscreen | |
Non-sterile | |
Digital User |