Learn More
Giese™ Agate Mortar with Pestle
Grind up solid chemicals and crush solids into smaller pieces with this agate mortar and pestle. Giese™ Agate Mortar with Pestle features a smooth, dense, and hard surface.
Brand:  Giese™ 600-M-115
4170.15 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11803360
  • 9865.00 NOK / Each
Only null left
Estimated Shipment: 19-09-2025
Description
  • Polished agate construction
  • For grinding and crushing
  • Durable
  • Available in different sizes
Specifications
Specifications
80 mL
Mortar and Pestle
Agate Stone
Documents
One moment while we fetch your results.
Certificates
View accessory products
One moment while we fetch your results.
Promotions
Promotions
Product Content Correction
Product Title
Giese™ Agate Mortar with Pestle > Capacity: 80 mL