Learn More
Giese™ Agate Mortar with Pestle
Grind up solid chemicals and crush solids into smaller pieces with this agate mortar and pestle. Giese™ Agate Mortar with Pestle features a smooth, dense, and hard surface.
Brand:  Giese™ 600M175
 View more versions of this product
Product Code. 11765146
  • 42620.00 NOK / Each
Only null left
Estimated Shipment: 07-10-2025
Description
  • Polished agate construction
  • For grinding and crushing
  • Durable
  • Available in different sizes
Specifications
Specifications
350 mL
Mortar and Pestle
Agate Stone
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Giese™ Agate Mortar with Pestle > Capacity: 350 mL