Learn More
Agar (80-100 Mesh), Fisher BioReagents
Brand: Fisher BioReagents BP2641-500
2164.27 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Solid | |
Agar | |
Poly Bottle | |
500 g |
Product Content Correction
Product Title
Agar (80-100 Mesh), Fisher BioReagents > Packaging: Poly Bottle; Quantity: 500g