Learn More
Eppendorf™ Aerosol-Tight Caps
Use these replacement lids with a variety of Eppendorf™ buckets.
Brand: Eppendorf™ 5810710006
1669.15 NOK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
This item is not returnable.
View return policy
Description
- Set of 2 caps
- Aerosol tight
Specifications
Cap | |
250mL Rectangular Bucket |
Product Content Correction
Product Title
Eppendorf™ Aerosol-Tight Caps > For 250mL Rectangular Bucket